missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)pyridine-5-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. T31881G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T31881G Supplier TCI America Supplier No. T31881G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 868662-36-6
Molecular Formula C6H5BF3NO2
Molecular Weight (g/mol) 190.916
MDL Number MFCD07375381
InChI Key BNTIPMNMTIAWIW-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyridine-5-boronic acid, 6-trifluoromethyl pyridin-3-yl boronic acid, 2-trifluoromethyl-5-pyridineboric acid, 2-trifluoromethyl pyridin-5-ylboronic acid, 6-trifluoromethyl pyridine-3-boronic acid, 6-trifluoromethyl pyridin-3-ylboronic acid, 2-trifluoromethylpyridine-5-boronic acid, 2-trifluoromethyl-5-pyridineboronic acid, 6-trifluoromethyl-3-pyridylboronic acid
PubChem CID 25134278
IUPAC Name [6-(trifluoromethyl)pyridin-3-yl]boronic acid
SMILES B(C1=CN=C(C=C1)C(F)(F)F)(O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 190.92
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethyl)pyridine-5-boronic Acid (contains varying amounts of Anhydride)