missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 61655-67-2 |
|---|---|
| Molecular Formula | C5H3F3N2 |
| Molecular Weight (g/mol) | 148.09 |
| MDL Number | MFCD06797734 |
| InChI Key | CEAMSISEJZMQEP-UHFFFAOYSA-N |
| Synonym | 2-trifluoromethyl pyrazine, 2-trifluoromethyl-pyrazine, pyrazine, trifluoromethyl, trifluoromethylpyrazine, pyrazine, 2-trifluoromethyl |
| PubChem CID | 17888936 |
| IUPAC Name | 2-(trifluoromethyl)pyrazine |
| SMILES | C1=CN=C(C=N1)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC432610050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 61655-67-2 | |
| 148.09 | |
| CEAMSISEJZMQEP-UHFFFAOYSA-N | |
| 17888936 | |
| C1=CN=C(C=N1)C(F)(F)F |
| C5H3F3N2 | |
| MFCD06797734 | |
| 2-trifluoromethyl pyrazine, 2-trifluoromethyl-pyrazine, pyrazine, trifluoromethyl, trifluoromethylpyrazine, pyrazine, 2-trifluoromethyl | |
| 2-(trifluoromethyl)pyrazine |
Specifications
| 61655-67-2 | |
| 100.0 | |
| Authentic | |
| C5H3F3N2 | |
| 2-trifluoromethyl pyrazine, 2-trifluoromethyl-pyrazine, pyrazine, trifluoromethyl, trifluoromethylpyrazine, pyrazine, 2-trifluoromethyl | |
| C1=CN=C(C=N1)C(F)(F)F | |
| 148.09 | |
| 148.09 | |
| 2-(Trifluoromethyl)pyrazine |
| 96.0 | |
| 116.0°C to 118.0°C | |
| 96% min. (GC) | |
| MFCD06797734 | |
| CEAMSISEJZMQEP-UHFFFAOYSA-N | |
| 2-(trifluoromethyl)pyrazine | |
| 17888936 | |
| 97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
GHS Signal Word: Warning
RUO – Research Use Only