missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)pyrazine, 97%

Catalog No. AAH3333803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3333803 1 g
AAH3333806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3333803 Supplier Thermo Scientific Chemicals Supplier No. H3333803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 61655-67-2 | C5H3F3N2 | 148.088 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61655-67-2
Molecular Formula C5H3F3N2
Molecular Weight (g/mol) 148.088
MDL Number MFCD06797734
InChI Key CEAMSISEJZMQEP-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyrazine, 2-trifluoromethyl-pyrazine, pyrazine, trifluoromethyl, trifluoromethylpyrazine, pyrazine, 2-trifluoromethyl
PubChem CID 17888936
IUPAC Name 2-(trifluoromethyl)pyrazine
SMILES C1=CN=C(C=N1)C(F)(F)F

Specifications

Boiling Point 116°C to 118°C
Quantity 1 g
UN Number UN2810
Formula Weight 148.09
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)pyrazine
Hazard Category H226-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only