missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)nicotinic Acid 97.0+%, TCI America™

Catalog No. T31321G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31321G 1 g
T31325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31321G Supplier TCI America Supplier No. T31321G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 131747-43-8 | C7H4F3NO2 | 191.11 g/mol

Chemical Identifiers

CAS 131747-43-8
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.11
MDL Number MFCD01862052
InChI Key BFROETNLEIAWNO-UHFFFAOYSA-N
Synonym 2-trifluoromethyl nicotinic acid, 2-trifluoromethylnicotinic acid, 2-trifluoromethyl pyridine-3-carboxylic acid, 2-trifluoromethyl-nicotinic acid, 3-pyridinecarboxylic acid, 2-trifluoromethyl, 2-trifluoromethyl-3-pyridinecarboxylic acid, pubchem12728, acmc-1c9gb, trifluoromethylnicotinic acid
PubChem CID 2777547
IUPAC Name 2-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES OC(=O)C1=C(N=CC=C1)C(F)(F)F

Specifications

Melting Point 187°C
Color White
Quantity 1 g
Formula Weight 191.11
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethyl)nicotinic Acid