missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)cyclohexanone, 97%

Catalog No. p-7032086
Change view
Click to view available options
Quantity:
250 mg
1 g
Catalog No. Quantity
AAH33423MD 250 mg
AAH3342303 1 g
2 options

Catalog No. AAH33423MD

Supplier: Thermo Scientific Chemicals H33423MD

Only null left

CAS: 56734-74-8 | C7H9F3O | 166.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56734-74-8
Molecular Formula C7H9F3O
Molecular Weight (g/mol) 166.14
MDL Number MFCD00102125
InChI Key RZSQHJQIBOWYSQ-UHFFFAOYNA-N
Synonym 2-trifluoromethyl cyclohexanone, 2-trifluoromethylcyclohexanone, 2-trifluoromethyl cyclohexan-1-one, trifluoromethylcyclohexanone, 2-trifluoromethyl-cyclohexan-1-one
PubChem CID 2777488
IUPAC Name 2-(trifluoromethyl)cyclohexan-1-one
SMILES FC(F)(F)C1CCCCC1=O

Specifications

Boiling Point 71°C to 73°C (12 mmHg)
Quantity 250 mg
UN Number UN1224
Formula Weight 166.14
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)cyclohexanone
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only