missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)cinnamic Acid 98.0+%, TCI America™

Catalog No. p-7137314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26085G 5 g
T260825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26085G Supplier TCI America Supplier No. T26085G

Chemical Identifiers

CAS 2062-25-1
Molecular Formula C10H7F3O2
Molecular Weight (g/mol) 216.16
MDL Number MFCD00004381
InChI Key AMVYAIXPAGBXOM-AATRIKPKSA-N
Synonym 2-trifluoromethyl cinnamic acid, o-trifluoromethyl cinnamic acid, 2-trifluoromethylcinnamic acid, e-3-2-trifluoromethyl phenyl acrylic acid, 2e-3-2-trifluoromethyl phenyl prop-2-enoic acid, 3-2-trifluoromethyl phenyl acrylic acid, 2e-3-2-trifluoroformyl phenyl acrylic acid, e-2-trifluoromethyl cinnamic acid, 3-2-trifluoromethyl-phenyl-acrylic acid
PubChem CID 719625
ChEBI CHEBI:60703
IUPAC Name (2E)-3-[2-(trifluoromethyl)phenyl]prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=CC=C1C(F)(F)F

Specifications

Melting Point 204°C
Color White-Yellow
Quantity 5 g
Formula Weight 216.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethyl)cinnamic Acid