missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzoyl chloride, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 312930050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC312930050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

312-94-7
208.56
MXIUWSYTQJLIKE-UHFFFAOYSA-N
67561
FC(F)(F)C1=CC=CC=C1C(Cl)=O
C8H4ClF3O
MFCD00000667
2-trifluoromethyl benzoyl chloride, o-trifluoromethylbenzoyl chloride, alpha,alpha,alpha-trifluoro-o-toluoyl chloride, 2-trifluoromethyl benzoylchloride, unii-d01d0b64eq, otf-boc, 2-trifluoromethyl-benzoyl chloride, o-trifluoromethyl benzoyl chloride
2-(trifluoromethyl)benzoyl chloride

Specifications

2-(Trifluoromethyl)benzoyl chloride
97.5
1.4100g/mL
95°C
97.5% min. (GC)
C8H4ClF3O
CF3C6H4COCl
MFCD00000667
2-trifluoromethyl benzoyl chloride, o-trifluoromethylbenzoyl chloride, alpha,alpha,alpha-trifluoro-o-toluoyl chloride, 2-trifluoromethyl benzoylchloride, unii-d01d0b64eq, otf-boc, 2-trifluoromethyl-benzoyl chloride, o-trifluoromethyl benzoyl chloride
MXIUWSYTQJLIKE-UHFFFAOYSA-N
2-(trifluoromethyl)benzoyl chloride
67561
98%
312-94-7
100.0
84°C to 85°C (16.0mmHg)
Authentic
Glass bottle
1.4780 to 1.481
5g
1.41
Solubility in water: decomposes
FC(F)(F)C1=CC=CC=C1C(Cl)=O
208.56
208.57
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-233-6

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...