missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzenethiol, ≥95%, Thermo Scientific™

Catalog No. p-2695306
Change view
Click to view available options
Quantity:
10g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
TL00684EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TL00684EA Supplier Thermo Scientific Chemicals Supplier No. TL00684EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13333-97-6
Molecular Formula C7H5F3S
Molecular Weight (g/mol) 178.172
MDL Number MFCD00068896
InChI Key XVGQHRKNXSUPEF-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzenethiol, 2-trifluoromethyl thiophenol, 2-trifluoromethylthiophenol, 2-trifluoromethyl benzene-1-thiol, 2-mercaptobenzotrifluoride, pubchem6860, acmc-209brx, trifluoromethyl thiophenol, 2trifluoromethylbenzenethiol
PubChem CID 2777889
IUPAC Name 2-(trifluoromethyl)benzenethiol
SMILES C1=CC=C(C(=C1)C(F)(F)F)S

Specifications

CAS Min % 95.0
Density 1.35g/mL
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula CF3C6H4SH
Quantity 10g
Formula Weight 178.18
Percent Purity 95%
Chemical Name or Material 2-(Trifluoromethyl)benzenethiol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.