missing translation for 'onlineSavingsMsg'
Learn More

2-Trifluoromethyl-2-propanol 97.0+%, TCI America™

Catalog No. T17105G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T17105G Supplier TCI America Supplier No. T17105G
Only null left

Chemical Identifiers

CAS 507-52-8
Molecular Formula C4H7F3O
Molecular Weight (g/mol) 128.09
MDL Number MFCD00014391
InChI Key OCGWWLDZAFOHGD-UHFFFAOYSA-N
Synonym 2-trifluoromethyl-2-propanol, 2-trifluoromethylpropanol-2, 2-propanol, 1,1,1-trifluoro-2-methyl, 2-trifluormethyl-2-propanol, acmc-1aya6, 2,2,2-trifluoro-tert-butanol, dimethyl trifluoromethyl methanol, 2-trifluoromethyl propan-2-ol, 1-methyl-1-trifluoromethyl-ethanol
PubChem CID 68182
IUPAC Name 1,1,1-trifluoro-2-methylpropan-2-ol
SMILES CC(C)(O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 82°C
Quantity 5 g
UN Number 1987
Formula Weight 128.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Trifluoromethyl-2-propanol