missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. T24131G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T24131G Supplier TCI America Supplier No. T24131G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 175676-65-0
Molecular Formula C7H6BF3O3
Molecular Weight (g/mol) 205.93
MDL Number MFCD01320763
InChI Key AIJCNTOYZPKURP-UHFFFAOYSA-N
PubChem CID 2777204
IUPAC Name [2-(trifluoromethoxy)phenyl]boronic acid
SMILES OB(O)C1=CC=CC=C1OC(F)(F)F

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 1 g
Synonym 2-trifluoromethoxy phenylboronic acid, 2-trifluoromethoxyphenylboronic acid, 2-trifluormethoxy phenylboronic acid, 2-trifluoromethoxy benzeneboronic acid, 2-trifluoromethoxy phenyl boronic acid, 2-trifluormethoxyphenylboronic acid, o-trifluoromethoxy phenylboronic acid, 2-trifluoromethoxy phenyl boranediol, boronic acid, 2-trifluoromethoxy phenyl
Formula Weight 205.93
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethoxy)phenylboronic Acid (contains varying amounts of Anhydride)