missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)benzaldehyde 95.0+%, TCI America™

Catalog No. T23025G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T23025G Supplier TCI America Supplier No. T23025G

Chemical Identifiers

CAS 94651-33-9
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.12
MDL Number MFCD00042405
InChI Key CPHXLFKIUVVIOQ-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy benzaldehyde, 4-trifluoromethoxy, o-trifluoromethoxy benzaldehyde, 2-trifluoromethoxy-benzaldehyde, pubchem1469, trifluoromethoxybenzaldehyde, acmc-209rt0, o-trifluoromethoxybenzaldehyde, trifluoromethoxy benzaldehyde
PubChem CID 2777192
IUPAC Name 2-(trifluoromethoxy)benzaldehyde
SMILES FC(F)(F)OC1=CC=CC=C1C=O

Specifications

Color Yellow
Boiling Point 76°C
Quantity 5 g
Formula Weight 190.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethoxy)benzaldehyde