Learn More
2-(Trifluoromethoxy)benzaldehyde, 96%
CAS: 94651-33-9 | C8H5F3O2 | 190.12 g/mol
Supplier: Thermo Scientific Chemicals B2084014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-(Trifluoromethoxy)benzaldehyde | |
| 1.33 | |
| 67°C (152°F) | |
| 1.455 | |
| 25 g | |
| Air Sensitive | |
| CPHXLFKIUVVIOQ-UHFFFAOYSA-N | |
| 2-(trifluoromethoxy)benzaldehyde | |
| 2777192 | |
| 96% |
| 94651-33-9 | |
| 77°C (20 mmHg) | |
| C8H5F3O2 | |
| MFCD00042405 | |
| 6137162 | |
| 2-trifluoromethoxy benzaldehyde, 4-trifluoromethoxy, o-trifluoromethoxy benzaldehyde, 2-trifluoromethoxy-benzaldehyde, pubchem1469, trifluoromethoxybenzaldehyde, acmc-209rt0, o-trifluoromethoxybenzaldehyde, trifluoromethoxy benzaldehyde | |
| FC(F)(F)OC1=CC=CC=C1C=O | |
| 190.12 | |
| 190.12 |
Chemical Identifiers
| 94651-33-9 | |
| 190.12 | |
| CPHXLFKIUVVIOQ-UHFFFAOYSA-N | |
| 2777192 | |
| FC(F)(F)OC1=CC=CC=C1C=O |
| C8H5F3O2 | |
| MFCD00042405 | |
| 2-trifluoromethoxy benzaldehyde, 4-trifluoromethoxy, o-trifluoromethoxy benzaldehyde, 2-trifluoromethoxy-benzaldehyde, pubchem1469, trifluoromethoxybenzaldehyde, acmc-209rt0, o-trifluoromethoxybenzaldehyde, trifluoromethoxy benzaldehyde | |
| 2-(trifluoromethoxy)benzaldehyde |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only