missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)benzaldehyde, 96%

Catalog No. AAB2084022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2084022 100 g
AAB2084006 5 g
AAB2084014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2084022 Supplier Thermo Scientific Chemicals Supplier No. B2084022

CAS: 94651-33-9 | C8H5F3O2 | 190.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 94651-33-9
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.12
MDL Number MFCD00042405
InChI Key CPHXLFKIUVVIOQ-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy benzaldehyde, 4-trifluoromethoxy, o-trifluoromethoxy benzaldehyde, 2-trifluoromethoxy-benzaldehyde, pubchem1469, trifluoromethoxybenzaldehyde, acmc-209rt0, o-trifluoromethoxybenzaldehyde, trifluoromethoxy benzaldehyde
PubChem CID 2777192
IUPAC Name 2-(trifluoromethoxy)benzaldehyde
SMILES FC(F)(F)OC1=CC=CC=C1C=O

Specifications

Density 1.33
Boiling Point 77°C (20 mmHg)
Flash Point 67°C (152°F)
Refractive Index 1.455
Quantity 100 g
Beilstein 6137162
Sensitivity Air Sensitive
Formula Weight 190.12
Percent Purity 96%
Chemical Name or Material 2-(Trifluoromethoxy)benzaldehyde
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only