missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 651-70-7 |
|---|---|
| Molecular Formula | C6H3F3OS |
| Molecular Weight (g/mol) | 180.15 |
| InChI Key | CZYKJGCKVBXLGF-UHFFFAOYSA-N |
| Synonym | 2-trifluoroacetyl thiophene, 2,2,2-trifluoro-1-thiophen-2-yl ethanone, ethanone, 2,2,2-trifluoro-1-2-thienyl, 2,2,2-trifluoro-1-2-thienyl ethanone, ethanone,2,2,2-trifluoro-1-2-thienyl, 2,2,2-trifluoro-1-thiophen-2-yl ethan-1-one, 2,2,2-trifluoro-1-2-thienyl ethan-1-one, acmc-20aolw, trifluoroacetylthiophene, 2-trifluoroacetylthiophene |
| PubChem CID | 136464 |
| IUPAC Name | 2,2,2-trifluoro-1-thiophen-2-ylethanone |
| SMILES | C1=CSC(=C1)C(=O)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC434700050
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 651-70-7 | |
| 180.15 | |
| 2-trifluoroacetyl thiophene, 2,2,2-trifluoro-1-thiophen-2-yl ethanone, ethanone, 2,2,2-trifluoro-1-2-thienyl, 2,2,2-trifluoro-1-2-thienyl ethanone, ethanone,2,2,2-trifluoro-1-2-thienyl, 2,2,2-trifluoro-1-thiophen-2-yl ethan-1-one, 2,2,2-trifluoro-1-2-thienyl ethan-1-one, acmc-20aolw, trifluoroacetylthiophene, 2-trifluoroacetylthiophene | |
| 2,2,2-trifluoro-1-thiophen-2-ylethanone |
| C6H3F3OS | |
| CZYKJGCKVBXLGF-UHFFFAOYSA-N | |
| 136464 | |
| C1=CSC(=C1)C(=O)C(F)(F)F |
Specifications
| 651-70-7 | |
| 100.0 | |
| 59°C | |
| 97.5% min. (GC) | |
| 2-trifluoroacetyl thiophene, 2,2,2-trifluoro-1-thiophen-2-yl ethanone, ethanone, 2,2,2-trifluoro-1-2-thienyl, 2,2,2-trifluoro-1-2-thienyl ethanone, ethanone,2,2,2-trifluoro-1-2-thienyl, 2,2,2-trifluoro-1-thiophen-2-yl ethan-1-one, 2,2,2-trifluoro-1-2-thienyl ethan-1-one, acmc-20aolw, trifluoroacetylthiophene, 2-trifluoroacetylthiophene | |
| C1=CSC(=C1)C(=O)C(F)(F)F | |
| 180.15 | |
| 180.15 | |
| 2-(Trifluoroacetyl)thiophene |
| 97.5 | |
| 163.0°C to 164.0°C | |
| Authentic | |
| C6H3F3OS | |
| CZYKJGCKVBXLGF-UHFFFAOYSA-N | |
| 2,2,2-trifluoro-1-thiophen-2-ylethanone | |
| 136464 | |
| 98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapour.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
RUO – Research Use Only