missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoroacetyl)thiophene, 98%

Catalog No. aal1258506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1258506 5 g
AAL1258503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1258506 Supplier Thermo Scientific Chemicals Supplier No. L1258506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 651-70-7 | C6H3F3OS | 180.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 651-70-7
Molecular Formula C6H3F3OS
Molecular Weight (g/mol) 180.144
MDL Number MFCD00041427
InChI Key CZYKJGCKVBXLGF-UHFFFAOYSA-N
Synonym 2-trifluoroacetyl thiophene, 2,2,2-trifluoro-1-thiophen-2-yl ethanone, ethanone, 2,2,2-trifluoro-1-2-thienyl, 2,2,2-trifluoro-1-2-thienyl ethanone, ethanone,2,2,2-trifluoro-1-2-thienyl, 2,2,2-trifluoro-1-thiophen-2-yl ethan-1-one, 2,2,2-trifluoro-1-2-thienyl ethan-1-one, acmc-20aolw, trifluoroacetylthiophene, 2-trifluoroacetylthiophene
PubChem CID 136464
IUPAC Name 2,2,2-trifluoro-1-thiophen-2-ylethanone
SMILES C1=CSC(=C1)C(=O)C(F)(F)F

Specifications

Density 1.403
Boiling Point 163°C to 165°C
Flash Point 58°C (136°F)
Refractive Index 1.4845
Quantity 5 g
UN Number UN1993
Beilstein 1309208
Formula Weight 180.15
Percent Purity 98%
Chemical Name or Material 2-(Trifluoroacetyl)thiophene
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only