missing translation for 'onlineSavingsMsg'
Learn More

2-Trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 99%

CAS: 74291-57-9 | C11H9ClF3NO3S | 327.70 g/mol

$289.51 - $289.51

Chemical Identifiers

CAS 74291-57-9
Molecular Formula C11H9ClF3NO3S
Molecular Weight (g/mol) 327.70
MDL Number MFCD01075651
InChI Key SXIYEBVAQTUBOQ-UHFFFAOYSA-N
Synonym 2-2,2,2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 1,2,3,4-tetrahydro-2-trifluoroacetyl isoquinoline-7-sulfonyl chloride, 2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 2-2,2,2-trifluoroacetyl-3,4-dihydro-1h-isoquinoline-7-sulfonyl chloride, 1-7-chlorosulfonyl 2-1,2,3,4-tetrahydroisoquinolyl-2,2,2-trifluoroethan-1-one, 2-trifluoroacetyl-7-chlorosulfonyl-1,2,3,4-tetrahydroisoquinoline, 7-chlorosulfonyl-2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline, 1,2,3,4-tetrahydro-2-trifluoroacetyl isoquinoline-7-sulfone
PubChem CID 11324902
IUPAC Name 2-(2,2,2-trifluoroacetyl)-3,4-dihydro-1H-isoquinoline-7-sulfonyl chloride
SMILES FC(F)(F)C(=O)N1CCC2=C(C1)C=C(C=C2)S(Cl)(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1358706
View Documents
Thermo Scientific Chemicals
L1358706
5 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

74291-57-9
327.70
SXIYEBVAQTUBOQ-UHFFFAOYSA-N
11324902
FC(F)(F)C(=O)N1CCC2=C(C1)C=C(C=C2)S(Cl)(=O)=O
C11H9ClF3NO3S
MFCD01075651
2-2,2,2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 1,2,3,4-tetrahydro-2-trifluoroacetyl isoquinoline-7-sulfonyl chloride, 2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 2-2,2,2-trifluoroacetyl-3,4-dihydro-1h-isoquinoline-7-sulfonyl chloride, 1-7-chlorosulfonyl 2-1,2,3,4-tetrahydroisoquinolyl-2,2,2-trifluoroethan-1-one, 2-trifluoroacetyl-7-chlorosulfonyl-1,2,3,4-tetrahydroisoquinoline, 7-chlorosulfonyl-2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline, 1,2,3,4-tetrahydro-2-trifluoroacetyl isoquinoline-7-sulfone
2-(2,2,2-trifluoroacetyl)-3,4-dihydro-1H-isoquinoline-7-sulfonyl chloride

Specifications

74291-57-9
C11H9ClF3NO3S
5 g
5603124
2-2,2,2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 1,2,3,4-tetrahydro-2-trifluoroacetyl isoquinoline-7-sulfonyl chloride, 2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 2-2,2,2-trifluoroacetyl-3,4-dihydro-1h-isoquinoline-7-sulfonyl chloride, 1-7-chlorosulfonyl 2-1,2,3,4-tetrahydroisoquinolyl-2,2,2-trifluoroethan-1-one, 2-trifluoroacetyl-7-chlorosulfonyl-1,2,3,4-tetrahydroisoquinoline, 7-chlorosulfonyl-2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline, 1,2,3,4-tetrahydro-2-trifluoroacetyl isoquinoline-7-sulfone
FC(F)(F)C(=O)N1CCC2=C(C1)C=C(C=C2)S(Cl)(=O)=O
327.70
327.71
2-Trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride
102°C to 106°C
MFCD01075651
UN3261
Moisture sensitive
SXIYEBVAQTUBOQ-UHFFFAOYSA-N
2-(2,2,2-trifluoroacetyl)-3,4-dihydro-1H-isoquinoline-7-sulfonyl chloride
11324902
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only