missing translation for 'onlineSavingsMsg'
Learn More

2-Tridecanol 98.0+%, TCI America™

Catalog No. T06335G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T06335G 5 g
T063325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T06335G Supplier TCI America Supplier No. T06335G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1653-31-2
Molecular Formula C13H28O
Molecular Weight (g/mol) 200.37
MDL Number MFCD00037601
InChI Key HKOLRKVMHVYNGG-UHFFFAOYNA-N
Synonym 2-tridecanol, 2-tridecyl alcohol, 2-hydroxytridecane, methylundecylcarbinol, 2-tridecanol, s, alcohols, c11-15-secondary, linear secondary c11-c15 alcohol, acmc-20m48e, acmc-1c00w
PubChem CID 15449
IUPAC Name tridecan-2-ol
SMILES CCCCCCCCCCCC(C)O

Specifications

Color Colorless
Boiling Point 265°C
Quantity 5 g
Formula Weight 200.37
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Tridecanol
EINECSNumber (2)-0217&(2)-3704
TSCA No