missing translation for 'onlineSavingsMsg'
Learn More

2-(Trichloroacetyl)pyrrole, 99+%

Catalog No. AAH6371806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6371806 5 g
AAH6371814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6371806 Supplier Thermo Scientific Chemicals Supplier No. H6371806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 35302-72-8 | C6H4Cl3NO | 212.454 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35302-72-8
Molecular Formula C6H4Cl3NO
Molecular Weight (g/mol) 212.454
MDL Number MFCD00128757
InChI Key BBFDGMDENAEMKF-UHFFFAOYSA-N
Synonym 2-trichloroacetyl pyrrole, 2,2,2-trichloro-1-1h-pyrrol-2-yl ethanone, 2,2,2-trichloro-1-1h-pyrrol-2-yl ethan-1-one, 2-trichloroacetyl-1h-pyrrole, 2-trichloroacetylpyrrole, 2,2,2-trichloro-1-1h-pyrrol-2-yl-ethanone, ethanone, 2,2,2-trichloro-1-1h-pyrrol-2-yl, 2,2,2-trichloro-1-pyrrol-2-ylethan-1-one, trichloroacetyl pyrrole
PubChem CID 321487
IUPAC Name 2,2,2-trichloro-1-(1H-pyrrol-2-yl)ethanone
SMILES C1=CNC(=C1)C(=O)C(Cl)(Cl)Cl

Specifications

Melting Point 72°C to 74°C
Color Gray
Quantity 5 g
Beilstein 135479
Formula Weight 212.46
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material 2-(Trichloroacetyl)pyrrole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only