missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Tri-n-butylstannyl)thiophene, 97%
Description
2-(Tri-n-butylstannyl)thiophene derivative is employed in Stille reaction involving efficient carbon-carbon bond formation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Tri-n-butylstannyl)thiophene derivative is employed in Stille reaction involving efficient carbon-carbon bond formation.
Notes
Incompatible with strong oxidizing agents.
2-(Tri-n-butylstannyl)thiophene derivative is employed in Stille reaction involving efficient carbon-carbon bond formation.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Color | Colorless to Yellow |
| Density | 1.175 g/mL |
| Boiling Point | 155°C (0.1 mmHg) |
| Refractive Index | 1.518 |
| Quantity | 5 mL |
| UN Number | UN2788 |
| Formula Weight | 373.16 |
| Percent Purity | 97% |
| Physical Form | Liquid |
| Chemical Name or Material | 2-(Tri-n-butylstannyl)thiophene |
Safety and Handling
| Hazard Category | H301-H312-H315-H319-H372 |
| Hazard Statement | GHS H Statement H301-H372-H312-H315-H319 Toxic if swallowed. Causes damage to organs through prolonged or repeated exposure. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only