Learn More
2-(Tri-n-butylstannyl)pyridine, tech. 80%
CAS: 17997-47-6 | C17H31NSn | 368.152 g/mol
Supplier: Thermo Scientific Chemicals B2491106
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-(Tri-n-butylstannyl)pyridine | |
| 1.15 | |
| C17H31NSn | |
| MFCD00052052 | |
| UN2788 | |
| 2-tributylstannyl pyridine, 2-tri-n-butylstannylpyridine, 2-tributylstannylpyridine, tributyl 2-pyridyl tin, 2-pyridyltributyltin, 2-1,1,1-tributylstannyl pyridine, 2-pyridyltri-n-butyltin, 2-tributylstannyl-pyridine, tri-n-butyl 2-pyridyl tin | |
| CCCC[Sn](CCCC)(CCCC)C1=CC=CC=N1 | |
| 368.152 | |
| 368.15 | |
| Technical |
| 17997-47-6 | |
| 130°C to 132°C (0.8 mmHg) | |
| 1.513 | |
| 5 g | |
| 479329 | |
| GYUURHMITDQTRU-UHFFFAOYSA-N | |
| tributyl(pyridin-2-yl)stannane | |
| 607784 | |
| 80% |
Chemical Identifiers
| 17997-47-6 | |
| 368.152 | |
| GYUURHMITDQTRU-UHFFFAOYSA-N | |
| 607784 | |
| CCCC[Sn](CCCC)(CCCC)C1=CC=CC=N1 |
| C17H31NSn | |
| MFCD00052052 | |
| 2-tributylstannyl pyridine, 2-tri-n-butylstannylpyridine, 2-tributylstannylpyridine, tributyl 2-pyridyl tin, 2-pyridyltributyltin, 2-1,1,1-tributylstannyl pyridine, 2-pyridyltri-n-butyltin, 2-tributylstannyl-pyridine, tri-n-butyl 2-pyridyl tin | |
| tributyl(pyridin-2-yl)stannane |
Safety and Handling
GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280i-P281-P301+P310-P303+P361+P353-P305+P351+P338-P308+P313-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H301-H312-H315-H319-H360FD-H372
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only