missing translation for 'onlineSavingsMsg'
Learn More

2-(Tri-n-butylstannyl)pyridine, tech. 80%

Catalog No. AAB2491103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2491103 1 g
AAB2491106 5 g
AAB2491114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAB2491103 Supplier Thermo Scientific Chemicals Supplier No. B2491103
Get Up to $1,800 in Chemicals with Purchase
View all promotions
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 17997-47-6 | C17H31NSn | 368.152 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17997-47-6
Molecular Formula C17H31NSn
Molecular Weight (g/mol) 368.152
MDL Number MFCD00052052
InChI Key GYUURHMITDQTRU-UHFFFAOYSA-N
Synonym 2-tributylstannyl pyridine, 2-tri-n-butylstannylpyridine, 2-tributylstannylpyridine, tributyl 2-pyridyl tin, 2-pyridyltributyltin, 2-1,1,1-tributylstannyl pyridine, 2-pyridyltri-n-butyltin, 2-tributylstannyl-pyridine, tri-n-butyl 2-pyridyl tin
PubChem CID 607784
IUPAC Name tributyl(pyridin-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CC=N1

Specifications

Density 1.15
Boiling Point 130°C to 132°C (0.8 mmHg)
Refractive Index 1.513
Quantity 1 g
UN Number UN2788
Beilstein 479329
Formula Weight 368.15
Percent Purity 80%
Grade Technical
Chemical Name or Material 2-(Tri-n-butylstannyl)pyridine
Hazard Category H226-H301-H312-H315-H319-H360FD-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280i-P281-P301+P310-P303+P361+P353-P305+P351+P338-P308+P313-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only