missing translation for 'onlineSavingsMsg'
Learn More

2-(Tri-n-butylstannyl)furan, 97%

Catalog No. p-7052674
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55259AA 5 mL
AAH55259AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAH55259AA Supplier Thermo Scientific Chemicals Supplier No. H55259AA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 118486-94-5 | C16H30OSn | 357.125 g/mol

It is an agrochemical, organic and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an agrochemical, organic and pharmaceutical intermediates.

Notes
Store at room temperature. Light sensitive.

Chemical Identifiers

CAS 118486-94-5
Molecular Formula C16H30OSn
Molecular Weight (g/mol) 357.125
MDL Number MFCD00192512
InChI Key SANWDQJIWZEKOD-UHFFFAOYSA-N
Synonym 2-tributylstannyl furan, tributyl furan-2-yl stannane, 2-tributylstannylfuran, tributyl 2-furyl tin, tributyl 2-furyl stannane, 2-tri-n-butylstannyl furan, stannane,tributyl-2-furanyl, stannane, tributyl-2-furanyl, 2-furyltributyltin, tributylstannylfuran
PubChem CID 4393688
IUPAC Name tributyl(furan-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CO1

Specifications

Color Colorless to Yellow
Density 1.134 g/mL
Boiling Point 90°C (0.1 mmHg)
Refractive Index 1.494
Quantity 5 mL
UN Number UN2788
Formula Weight 357.1
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-(Tri-n-butylstannyl)furan
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only