missing translation for 'onlineSavingsMsg'
Learn More

2-(Tri-n-butylstannyl)benzo[b]thiophene, 95%, Thermo Scientific™

Catalog No. p-7052693
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55348MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH55348MD Supplier Thermo Scientific Chemicals Supplier No. H55348MD
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 148961-88-0
Molecular Formula C20H32SSn
Molecular Weight (g/mol) 423.246
MDL Number MFCD01318936
InChI Key UJPVMOUQRNYNSA-UHFFFAOYSA-N
Synonym 2-tributylstannylbenzo b thiophene, benzo b thiophen-2-yltributylstannane, stannane,benzo b thien-2-yltributyl, 2-tri-n-butylstannyl benzo b thiophene, acmc-20anmg, 2-benzothienyl tributyltin, benzo b thien-2-yltributylstannane, benzo b thien-2-yltributyl-stannane, 1-benzothien-2-yl tributyl stannane, benzo b thiophen-2-yl-tributyltin
PubChem CID 2735461
IUPAC Name 1-benzothiophen-2-yl(tributyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC2=CC=CC=C2S1

Specifications

Color Colorless
Density 1.184 g/mL
Refractive Index 1.564
Quantity 250 mg
UN Number UN2788
Formula Weight 423.22
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-(Tri-n-butylstannyl)benzo[b]thiophene
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures