missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenemethanethiol 95.0+%, TCI America™

Catalog No. p-7137316
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26131G 1 g
T26135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26131G Supplier TCI America Supplier No. T26131G
Only null left

Chemical Identifiers

CAS 6258-63-5
Molecular Formula C5H6S2
Molecular Weight (g/mol) 130.223
MDL Number MFCD00107135
InChI Key GCZQHDFWKVMZOE-UHFFFAOYSA-N
Synonym 2-thiophenemethanethiol, thenyl mercaptan, 2-thenylmercaptan, 2-thienylmethanethiol, thiophene-2-methanethiol, 2-thenylthiol, thiophen-2-yl-methanethiol, unii-9jml279xs6, 2-thenyl mercaptan, 2-mercaptomethyl thiophene
PubChem CID 80408
IUPAC Name thiophen-2-ylmethanethiol
SMILES C1=CSC(=C1)CS

Specifications

Color Yellow
Boiling Point 86°C
Quantity 1 g
Formula Weight 130.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Thiophenemethanethiol