missing translation for 'onlineSavingsMsg'
Learn More

2-Thiopheneethanol, 98%

Catalog No. AAB2156222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2156222 100 g
AAB2156206 5 g
AAB2156214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2156222 Supplier Thermo Scientific Chemicals Supplier No. B2156222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5402-55-1 | C6H8OS | 128.19 g/mol

2-Thiopheneethanol, is used as the antithrombotics thiophene chlorine match piperidine intermediates. It is also used as a ticlopidine Intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Thiopheneethanol, is used as the antithrombotics thiophene chlorine match piperidine intermediates. It is also used as a ticlopidine Intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents.

Chemical Identifiers

CAS 5402-55-1
Molecular Formula C6H8OS
Molecular Weight (g/mol) 128.19
MDL Number MFCD00005462
InChI Key VMJOFTHFJMLIKL-UHFFFAOYSA-N
Synonym 2-thiopheneethanol, 2-2-thienyl ethanol, thiophene-2-ethanol, 2-thiophen-2-yl ethanol, 2-2-hydroxyethyl thiophene, 2-thiophen-2-yl ethan-1-ol, ethanol, 2-2-thienyl, 2-thiophenethanol, 2-2-thienyl-ethanol, 2-thienylethanol
PubChem CID 79400
IUPAC Name 2-thiophen-2-ylethanol
SMILES OCCC1=CC=CS1

Specifications

Density 1.15
Boiling Point 107°C to 108°C (12 mmHg)
Flash Point 101°C (213°F)
Refractive Index 1.551
Quantity 100 g
Beilstein 106985
Solubility Information Slightly soluble in water.
Formula Weight 128.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Thiopheneethanol
Show More Show Less
EINECSNumber 226-452-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only