missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Thiopheneethanol 98.0+%, TCI America™
$75.08 - $75.08
Chemical Identifiers
| CAS | 5402-55-1 |
|---|---|
| Molecular Formula | C6H8OS |
| Molecular Weight (g/mol) | 128.19 |
| MDL Number | MFCD00005462 |
| InChI Key | VMJOFTHFJMLIKL-UHFFFAOYSA-N |
| Synonym | 2-thiopheneethanol, 2-2-thienyl ethanol, thiophene-2-ethanol, 2-thiophen-2-yl ethanol, 2-2-hydroxyethyl thiophene, 2-thiophen-2-yl ethan-1-ol, ethanol, 2-2-thienyl, 2-thiophenethanol, 2-2-thienyl-ethanol, 2-thienylethanol |
| PubChem CID | 79400 |
| IUPAC Name | 2-(thiophen-2-yl)ethan-1-ol |
| SMILES | OCCC1=CC=CS1 |
Chemical Identifiers
| 5402-55-1 | |
| 128.19 | |
| VMJOFTHFJMLIKL-UHFFFAOYSA-N | |
| 79400 | |
| OCCC1=CC=CS1 |
| C6H8OS | |
| MFCD00005462 | |
| 2-thiopheneethanol, 2-2-thienyl ethanol, thiophene-2-ethanol, 2-thiophen-2-yl ethanol, 2-2-hydroxyethyl thiophene, 2-thiophen-2-yl ethan-1-ol, ethanol, 2-2-thienyl, 2-thiophenethanol, 2-2-thienyl-ethanol, 2-thienylethanol | |
| 2-(thiophen-2-yl)ethan-1-ol |
Specifications
| 5402-55-1 | |
| 115°C | |
| MFCD00005462 | |
| 2-thiopheneethanol, 2-2-thienyl ethanol, thiophene-2-ethanol, 2-thiophen-2-yl ethanol, 2-2-hydroxyethyl thiophene, 2-thiophen-2-yl ethan-1-ol, ethanol, 2-2-thienyl, 2-thiophenethanol, 2-2-thienyl-ethanol, 2-thienylethanol | |
| OCCC1=CC=CS1 | |
| 128.19 | |
| 128.19 | |
| Liquid |
| Yellow | |
| C6H8OS | |
| 25 mL | |
| VMJOFTHFJMLIKL-UHFFFAOYSA-N | |
| 2-(thiophen-2-yl)ethan-1-ol | |
| 79400 | |
| ≥98.0% (GC) | |
| 2-Thiopheneethanol |
Safety and Handling
TSCA : Yes