missing translation for 'onlineSavingsMsg'
Learn More

2-Thiopheneethanol 98.0+%, TCI America™

Catalog No. T102125ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T102125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T102125ML Supplier TCI America Supplier No. T102125ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5402-55-1
Molecular Formula C6H8OS
Molecular Weight (g/mol) 128.19
MDL Number MFCD00005462
InChI Key VMJOFTHFJMLIKL-UHFFFAOYSA-N
Synonym 2-thiopheneethanol, 2-2-thienyl ethanol, thiophene-2-ethanol, 2-thiophen-2-yl ethanol, 2-2-hydroxyethyl thiophene, 2-thiophen-2-yl ethan-1-ol, ethanol, 2-2-thienyl, 2-thiophenethanol, 2-2-thienyl-ethanol, 2-thienylethanol
PubChem CID 79400
IUPAC Name 2-(thiophen-2-yl)ethan-1-ol
SMILES OCCC1=CC=CS1

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 mL
Formula Weight 128.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Thiopheneethanol