missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenecarboxylic Hydrazide 98.0+%, TCI America™

Catalog No. T114010G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T114010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T114010G Supplier TCI America Supplier No. T114010G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2361-27-5
Molecular Formula C5H6N2OS
Molecular Weight (g/mol) 142.176
MDL Number MFCD00005435
InChI Key SOGBOGBTIKMGFS-UHFFFAOYSA-N
Synonym 2-thiophenecarboxylic acid hydrazide, thiophene-2-carboxylic acid hydrazide, 2-thenoylhydrazine, 2-thiophenecarboxylic hydrazide, 2-thiophenecarbohydrazide, thiophene-2-carboxylic hydrazide, 2-thiophenecarboxylic acid, hydrazide, 2-thiophenecarbohydrazonic acid, 2-thienylcarboxylic acid hydrazide, 2-thiophene carboxylic hydrazide
PubChem CID 73098
IUPAC Name thiophene-2-carbohydrazide
SMILES C1=CSC(=C1)C(=O)NN

Specifications

Melting Point 139°C
Quantity 10 g
Formula Weight 142.18
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Thiophenecarboxylic Hydrazide