missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenecarboxylic Acid 98.0+%, TCI America™

Catalog No. T021825G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T021825G 25 g
T0218100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T021825G Supplier TCI America Supplier No. T021825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 527-72-0
Molecular Formula C5H4O2S
Molecular Weight (g/mol) 128.15
MDL Number MFCD00005437
InChI Key QERYCTSHXKAMIS-UHFFFAOYSA-N
Synonym 2-thiophenecarboxylic acid, 2-thenoic acid, 2-carboxythiophene, 2-thiophenic acid, 2-thienylcarboxylic acid, alpha-thiophenecarboxylic acid, 2-tca, thiophene-2-carboxylate, tenoic acid, .alpha.-thiophenecarboxylic acid
PubChem CID 10700
ChEBI CHEBI:71241
IUPAC Name thiophene-2-carboxylic acid
SMILES OC(=O)C1=CC=CS1

Specifications

Melting Point 128°C
Boiling Point 260°C
Quantity 25 g
Formula Weight 128.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Thiophenecarboxylic Acid
RTECSNumber XM8330200
TSCA Yes