missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 5813-89-8 |
|---|---|
| Molecular Formula | C5H5NOS |
| Molecular Weight (g/mol) | 127.17 |
| MDL Number | MFCD00010425 |
| InChI Key | DENPQNAWGQXKCU-UHFFFAOYSA-N |
| Synonym | 2-thiophenecarboxamide, 2-thienylamide, 2-thenamide, thiophen-2-carboxamide, 2-aminocarbonyl thiophene, 2-thienylamide french, thiophenecarboxamide, thiolamide, acmc-1ao1m, 5-18-06-00162 beilstein handbook reference |
| PubChem CID | 22063 |
| IUPAC Name | thiophene-2-carboxamide |
| SMILES | C1=CSC(=C1)C(=O)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC305420050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| C5H5NOS | |
| MFCD00010425 | |
| 2-thiophenecarboxamide, 2-thienylamide, 2-thenamide, thiophen-2-carboxamide, 2-aminocarbonyl thiophene, 2-thienylamide french, thiophenecarboxamide, thiolamide, acmc-1ao1m, 5-18-06-00162 beilstein handbook reference | |
| thiophene-2-carboxamide |
Specifications
| 5813-89-8 | |
| 100.0 | |
| 98.5% min. (GC) | |
| MFCD00010425 | |
| DENPQNAWGQXKCU-UHFFFAOYSA-N | |
| thiophene-2-carboxamide | |
| 22063 | |
| 99% |
| 98.5 | |
| Authentic | |
| C5H5NOS | |
| 2-thiophenecarboxamide, 2-thienylamide, 2-thenamide, thiophen-2-carboxamide, 2-aminocarbonyl thiophene, 2-thienylamide french, thiophenecarboxamide, thiolamide, acmc-1ao1m, 5-18-06-00162 beilstein handbook reference | |
| C1=CSC(=C1)C(=O)N | |
| 127.17 | |
| 127.17 | |
| 2-Thiophenecarboxamide |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
RUO – Research Use Only