missing translation for 'onlineSavingsMsg'
Learn More

2-Thiopheneacrylic Acid 98.0+%, TCI America™

Catalog No. T11385G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11385G 5 g
T113825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T11385G Supplier TCI America Supplier No. T11385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15690-25-2
Molecular Formula C7H6O2S
Molecular Weight (g/mol) 154.183
MDL Number MFCD00005455
InChI Key KKMZQOIASVGJQE-ONEGZZNKSA-N
Synonym 3-2-thienyl acrylic acid, 3-thiophen-2-yl acrylic acid, 2-thiopheneacrylic acid, 2e-3-thiophen-2-yl prop-2-enoic acid, 2e-3-2-thienyl acrylic acid, 2-thienylacrylic acid, e-3-thiophen-2-yl acrylic acid, 2-thiopheneacrylic acid, e, unii-ino5aty68d, 2-propenoic acid, 3-2-thienyl
PubChem CID 735981
IUPAC Name (E)-3-thiophen-2-ylprop-2-enoic acid
SMILES C1=CSC(=C1)C=CC(=O)O

Specifications

Melting Point 146°C
Quantity 5 g
Formula Weight 154.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Thiopheneacrylic Acid