missing translation for 'onlineSavingsMsg'
Learn More

2-Thiopheneacetic acid, 98%

CAS: 1918-77-0 | C6H6O2S | 142.172 g/mol

Supplier:  Thermo Scientific Chemicals A1039422

 View more versions of this product

Catalog No. AAA1039422

Only null left
Explore available promotions

Description

Description

2-Thiopheneacetic acid is used in the preparation of rosette-like nanoscale gold materials. Further, it is used as a pharmaceutical intermediate for cefaloridine and cefalotin sodium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Thiopheneacetic acid is used in the preparation of rosette-like nanoscale gold materials. Further, it is used as a pharmaceutical intermediate for cefaloridine and cefalotin sodium.

Solubility
Soluble in water, ethanol, ether and carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

2-Thiopheneacetic acid
61°C to 65°C
C6H6O2S
100 g
114551
Soluble in water,ethanol,ether and carbon tetrachloride.
C1=CSC(=C1)CC(=O)O
142.172
CHEBI:45807
98%
1918-77-0
160°C (22 mmHg)
MFCD00005458
UN3261
2-thiopheneacetic acid, 2-thienylacetic acid, thiopheneacetic acid, 2-thiophen-2-yl acetic acid, thiophene-2-acetic acid, 2-2-thienyl acetic acid, 2-thiophene acetic acid, thiophen-2-ylacetic acid, thien-2-yl acetic acid, 2-carboxymethyl thiophene
SMJRBWINMFUUDS-UHFFFAOYSA-N
2-thiophen-2-ylacetic acid
15970
142.18

Chemical Identifiers

1918-77-0
142.172
SMJRBWINMFUUDS-UHFFFAOYSA-N
15970
2-thiophen-2-ylacetic acid
C6H6O2S
MFCD00005458
2-thiopheneacetic acid, 2-thienylacetic acid, thiopheneacetic acid, 2-thiophen-2-yl acetic acid, thiophene-2-acetic acid, 2-2-thienyl acetic acid, 2-thiophene acetic acid, thiophen-2-ylacetic acid, thien-2-yl acetic acid, 2-carboxymethyl thiophene
CHEBI:45807
C1=CSC(=C1)CC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 217-639-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only