missing translation for 'onlineSavingsMsg'
Learn More

2-Thiomorpholinoisonicotinaldehyde, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC50504DA

 View more versions of this product

Catalog No. CC50504DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

2-Thiomorpholinoisonicotinaldehyde
96.5
97%
C10H12N2OS
1g
XESFZSHCANCGBY-UHFFFAOYSA-N
2-thiomorpholin-4-ylpyridine-4-carbaldehyde
24229507
97%
898289-24-2
100.0
Amber Glass Bottle
MFCD09702361
2-thiomorpholinoisonicotinaldehyde, 2-thiomorpholin-4-yl pyridine-4-carbaldehyde, 2-thiomorpholin-4-yl pyridine-4-carboxaldehyde, 4-pyridinecarboxaldehyde,2-4-thiomorpholinyl, 2-thiomorpholin-4-yl isonicotinaldehyde, 2-1,4-thiazaperhydroin-4-yl pyridine-4-carbaldehyde
O=CC1=CC(=NC=C1)N1CCSCC1
208.28
208.28

Chemical Identifiers

898289-24-2
208.28
XESFZSHCANCGBY-UHFFFAOYSA-N
24229507
O=CC1=CC(=NC=C1)N1CCSCC1
C10H12N2OS
MFCD09702361
2-thiomorpholinoisonicotinaldehyde, 2-thiomorpholin-4-yl pyridine-4-carbaldehyde, 2-thiomorpholin-4-yl pyridine-4-carboxaldehyde, 4-pyridinecarboxaldehyde,2-4-thiomorpholinyl, 2-thiomorpholin-4-yl isonicotinaldehyde, 2-1,4-thiazaperhydroin-4-yl pyridine-4-carbaldehyde
2-thiomorpholin-4-ylpyridine-4-carbaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions