missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Thiohydantoin, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 503-87-7 |
|---|---|
| Molecular Formula | C3H4N2OS |
| Molecular Weight (g/mol) | 116.14 |
| MDL Number | MFCD00005277 |
| InChI Key | UGWULZWUXSCWPX-UHFFFAOYSA-N |
| Synonym | 2-thiohydantoin, thiohydantoin, 4-imidazolidinone, 2-thioxo, 2-thioxoimidazolidin-4-one, 2-thioguidanthion, hydantoin, 2-thio, usaf be-25, 2-thioxo-4-imidazolidinone, 2-thioxo-1,3-diazolidin-4-one, glycine thiohydantoin |
| PubChem CID | 1274030 |
| IUPAC Name | 2-sulfanylideneimidazolidin-4-one |
| SMILES | O=C1CNC(=S)N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC138760050
|
Thermo Scientific Chemicals
138760050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC138761000
|
Thermo Scientific Chemicals
138761000 |
100g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 503-87-7 | |
| 116.14 | |
| UGWULZWUXSCWPX-UHFFFAOYSA-N | |
| 1274030 | |
| O=C1CNC(=S)N1 |
| C3H4N2OS | |
| MFCD00005277 | |
| 2-thiohydantoin, thiohydantoin, 4-imidazolidinone, 2-thioxo, 2-thioxoimidazolidin-4-one, 2-thioguidanthion, hydantoin, 2-thio, usaf be-25, 2-thioxo-4-imidazolidinone, 2-thioxo-1,3-diazolidin-4-one, glycine thiohydantoin | |
| 2-sulfanylideneimidazolidin-4-one |
Specifications
| 503-87-7 | |
| 97.5% min. (HPLC) | |
| C3H4N2OS | |
| 5g | |
| 2-thiohydantoin, thiohydantoin, 4-imidazolidinone, 2-thioxo, 2-thioxoimidazolidin-4-one, 2-thioguidanthion, hydantoin, 2-thio, usaf be-25, 2-thioxo-4-imidazolidinone, 2-thioxo-1,3-diazolidin-4-one, glycine thiohydantoin | |
| UGWULZWUXSCWPX-UHFFFAOYSA-N | |
| 2-sulfanylideneimidazolidin-4-one | |
| 1274030 | |
| 98% |
| Authentic | |
| Glass bottle | |
| MFCD00005277 | |
| 24,26 | |
| Solubility in water: slightly. | |
| O=C1CNC(=S)N1 | |
| 116.14 | |
| 116.14 | |
| 2-Thiohydantoin |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
GHS Signal Word: Warning
EINECSNumber : 207-977-4
RUO â Research Use Only