missing translation for 'onlineSavingsMsg'
Learn More

(2-Thien-2-ylphenyl)methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC35509DA

Quantity 1g

 View more versions of this product

Catalog No. CC35509DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

773872-97-2
190.26
IUULMJAEIKVTKZ-UHFFFAOYSA-N
2795559
OCC1=CC=CC=C1C1=CC=CS1
C11H10OS
MFCD06203081
2-thien-2-ylphenyl methanol, 2-thiophen-2-yl phenyl methanol, 2-thien-2-yl phenyl methanol, 2-thiophen-2-ylphenyl methanol, 2-2-thienyl phenyl methanol, benzenemethanol,2-2-thienyl, 2-2-thienyl phenyl methan-1-ol
[2-(thiophen-2-yl)phenyl]methanol

Specifications

(2-Thien-2-ylphenyl)methanol
96.5
97%
C11H10OS
MFCD06203081
IUULMJAEIKVTKZ-UHFFFAOYSA-N
[2-(thiophen-2-yl)phenyl]methanol
2795559
97%
773872-97-2
100.0
Amber glass bottle
1g
2-thien-2-ylphenyl methanol, 2-thiophen-2-yl phenyl methanol, 2-thien-2-yl phenyl methanol, 2-thiophen-2-ylphenyl methanol, 2-2-thienyl phenyl methanol, benzenemethanol,2-2-thienyl, 2-2-thienyl phenyl methan-1-ol
OCC1=CC=CC=C1C1=CC=CS1
190.26
190.27
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents

Promotions

Promotions