missing translation for 'onlineSavingsMsg'
Learn More

2-Thenylmercaptan, 98%

Catalog No. AC265110010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC265110010 1 g Glass bottle
AC265110050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC265110010 Supplier Thermo Scientific Chemicals Supplier No. 265110010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6258-63-5 | C5H6S2 | 130.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6258-63-5
Molecular Formula C5H6S2
Molecular Weight (g/mol) 130.22
MDL Number MFCD00107135
InChI Key GCZQHDFWKVMZOE-UHFFFAOYSA-N
Synonym 2-thiophenemethanethiol, thenyl mercaptan, 2-thenylmercaptan, 2-thienylmethanethiol, thiophene-2-methanethiol, 2-thenylthiol, thiophen-2-yl-methanethiol, unii-9jml279xs6, 2-thenyl mercaptan, 2-mercaptomethyl thiophene
PubChem CID 80408
IUPAC Name thiophen-2-ylmethanethiol
SMILES C1=CSC(=C1)CS

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2000g/mL
Boiling Point 90°C (15.0 mmHg)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.6020 to 1.604
Quantity 1 g
Specific Gravity 1.2
Solubility Information Solubility in water: slightly soluble
Formula Weight 130.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Thenylmercaptan
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 228-394-1

RUO – Research Use Only