missing translation for 'onlineSavingsMsg'
Learn More

2-(Tetrazol-5-yl)phenylboronic acid, 95%

Catalog No. AC445550010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445550010 1 g
AC445550050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC445550010 Supplier Thermo Scientific Chemicals Supplier No. 445550010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 155884-01-8 | C7H7BN4O2 | 189.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 155884-01-8
Molecular Formula C7H7BN4O2
Molecular Weight (g/mol) 189.97
MDL Number MFCD06796325
InChI Key GVRXWYFECKHTSJ-UHFFFAOYSA-N
Synonym 2-5-tetrazolyl phenylboronic acid, 2-1h-tetrazol-5-yl phenyl boronic acid, 2-tetrazol-5-yl phenylboronic acid, 2-2h-tetrazol-5-yl phenylboronic acid, 2-2h-tetrazol-5-yl-phenylboronic acid, 2-1h-tetrazol-5-yl phenylboronic acid, 2-2h-tetrazol-5-yl phenyl boronic acid, boronic acid, 2-1h-tetrazol-5-yl phenyl, boronic acid,b-2-2h-tetrazol-5-yl phenyl
PubChem CID 11171482
IUPAC Name [2-(2H-1,2,3,4-tetrazol-5-yl)phenyl]boronic acid
SMILES OB(O)C1=CC=CC=C1C1=NNN=N1

Specifications

Melting Point 148°C to 152°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 189.97
Percent Purity 95%
Chemical Name or Material 2-(Tetrazol-5-yl)phenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

RUO – Research Use Only