missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Hydrogen Chloride Methanol Solution, ∼1.25 M HCl (T), For GC Derivatization, MilliporeSigma™ Supelco™

Catalog No. p-7193393
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020464 50 mL
111019040 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020464 Supplier Merck Emd Millipore Supplier No. 1793550ML

Specifications

CAS 132228-87-6
Density 0.831 g/mL (at 20°C)
Linear Formula HCl · CH3OH
MDL Number MFCD00011324
Quantity 50 mL
Synonym Hydrochloric acid in methanol
Formula Weight 36.46
Grade For GC derivatization
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany