missing translation for 'onlineSavingsMsg'
Learn More

2-(Tetrahydropyran-4-yloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC58339CB

 View more versions of this product

Catalog No. CC58339CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

2-(Tetrahydropyran-4-yloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
96.5
97%
C16H24BNO4
250mg
KGCJDMWHIVJQGF-UHFFFAOYSA-N
2-(oxan-4-yloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
24229583
97%
910036-98-5
100.0
Amber Glass Bottle
MFCD09702390
2-tetrahydro-pyran-4-yloxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-tetrahydropyran-4-yloxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-tetrahydro-2h-pyran-4-yl oxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-oxan-4-yloxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-tetrahydro-2h-pyran-4-yloxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-oxan-4-yloxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pubchem20144, 2-tetrahydropyran-4-yloxy pyridine-5-boronic acid pinacol ester, 6-tetrahydropyran-4-yloxy pyridine-3-boronic acid pinacol ester, 6-tetrahydro-2h-pyran-4-yl oxy pyridin-3-yl boronic acid pinacol ester
CC1(C)OB(OC1(C)C)C1=CN=C(OC2CCOCC2)C=C1
305.18
305.18

Chemical Identifiers

910036-98-5
305.18
KGCJDMWHIVJQGF-UHFFFAOYSA-N
24229583
CC1(C)OB(OC1(C)C)C1=CN=C(OC2CCOCC2)C=C1
C16H24BNO4
MFCD09702390
2-tetrahydro-pyran-4-yloxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-tetrahydropyran-4-yloxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-tetrahydro-2h-pyran-4-yl oxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-oxan-4-yloxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-tetrahydro-2h-pyran-4-yloxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-oxan-4-yloxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pubchem20144, 2-tetrahydropyran-4-yloxy pyridine-5-boronic acid pinacol ester, 6-tetrahydropyran-4-yloxy pyridine-3-boronic acid pinacol ester, 6-tetrahydro-2h-pyran-4-yl oxy pyridin-3-yl boronic acid pinacol ester
2-(oxan-4-yloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions