missing translation for 'onlineSavingsMsg'
Learn More

2-Tetradecanone 97.0+%, TCI America™

Catalog No. T13141G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T13141G Supplier TCI America Supplier No. T13141G
Only null left

Chemical Identifiers

CAS 2345-27-9
Molecular Formula C14H28O
Molecular Weight (g/mol) 212.377
MDL Number MFCD00060927
InChI Key POQLVOYRGNFGRM-UHFFFAOYSA-N
Synonym Dodecyl Methyl Ketone
PubChem CID 75364
IUPAC Name tetradecan-2-one
SMILES CCCCCCCCCCCCC(=O)C

Specifications

Melting Point 35°C
Color White-Yellow
Boiling Point 148°C
Quantity 1 g
Formula Weight 212.38
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Tetradecanone