missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butylpropane-1,3-diol, 98%, Thermo Scientific Chemicals

Catalog No. AAL1692706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1692706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1692706 Supplier Thermo Scientific Chemicals Supplier No. L1692706
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2819-05-8
MDL Number MFCD01075745

Specifications

Melting Point 54°C to 56°C
Boiling Point 100°C to 102°C (2 mmHg)
Molecular Formula C10H16O4
Quantity 5 g
Beilstein 1900687
Synonym ethyl 4,4-diacetylbutyrate, 4-acetyl-5-oxohexanoic acid ethyl ester, 4,4-diacetylbutyric acid ethyl ester, ethyl 4-acetyl-5-oxohexanoate, 4-acetyl-5-oxo-hexanoic acid ethyl ester, hexanoic acid,4-acetyl-5-oxo-, ethyl ester, hexanoic acid, 4-acetyl-5-oxo-, ethyl ester
InChI Key CVPCFJRJUCNUJG-VQHVLOKHSA-N
SMILES CCOC(=O)CCC(C(=O)C)C(=O)C
IUPAC Name ethyl 4-acetyl-5-oxohexanoate
Molecular Weight (g/mol) 200.23
PubChem CID 2733934
Formula Weight 132.2
Percent Purity 98%
Chemical Name or Material 2-tert-Butylpropane-1,3-diol
Show More Show Less
EINECSNumber 000-000-0
RTECSNumber TY3540020
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only