missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine, 98%

Catalog No. p-4229132
Change view
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC439160010 1 mL Glass bottle
AC439160050 5 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC439160010 Supplier Thermo Scientific Chemicals Supplier No. 439160010

CAS: 98015-45-3 | C13H31N4P | 274.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98015-45-3
Molecular Formula C13H31N4P
Molecular Weight (g/mol) 274.39
InChI Key VSCBATMPTLKTOV-UHFFFAOYSA-N
Synonym bemp, bemp phosphazene, 2-tert-butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine, unii-df15j146qi, 2-tert-butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine, 2-tert-butylimino-n,n-diethyl-1,3-dimethyl-1,3,2??-diazaphosphinan-2-amine, 2-tert-butylimino-n,n-diethyl-1,3-dimethyl-1,3,2, 2-t-butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine, 2-tert-butylimino-2-diethylamino-1,3-dimethyl-1,3,2-diazaphosphorinane
PubChem CID 3513851
IUPAC Name 2-tert-butylimino-N,N-diethyl-1,3-dimethyl-1,3,2$l^{5}-diazaphosphinan-2-amine
SMILES CCN(CC)P1(=NC(C)(C)C)N(CCCN1C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9480g/mL
Boiling Point 74°C (0.1 mmHg)
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 mL
Specific Gravity 0.948
Formula Weight 274.39
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-tert-Butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

RUO – Research Use Only