missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butyl-2-[2-(4-chlorophenyl)ethyl]oxirane 96.0+%, TCI America™

Catalog No. B32905G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B32905G Supplier TCI America Supplier No. B32905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 80443-63-6
Quantity 5 g
Physical Form Clear Liquid at 20°C
Molecular Formula C14H19ClO
MDL Number MFCD01938345
InChI Key QLCGXXYDHCTVKP-UHFFFAOYSA-N
SMILES CC(C)(C)C1(CO1)CCC2=CC=C(C=C2)Cl
IUPAC Name 2-tert-butyl-2-[2-(4-chlorophenyl)ethyl]oxirane
Molecular Weight (g/mol) 238.755
PubChem CID 94684
Formula Weight 238.76
Percent Purity ≥96.0% (GC)
Chemical Name or Material 2-tert-Butyl-2-[2-(4-chlorophenyl)ethyl]oxirane
Show More Show Less
RTECSNumber RQ2850750
TSCA Yes