missing translation for 'onlineSavingsMsg'
Learn More

2-(tert-Butoxycarbonylamino)ethyl Bromide 98.0+%, TCI America™

Catalog No. B22891G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22891G 1 g
B22895G 5 g
B228925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B22891G Supplier TCI America Supplier No. B22891G

Chemical Identifiers

CAS 39684-80-5
Molecular Formula C7H14BrNO2
Molecular Weight (g/mol) 224.10
MDL Number MFCD02683428
InChI Key TZRQZPMQUXEZMC-UHFFFAOYSA-N
Synonym tert-butyl n-2-bromoethyl carbamate, 2-boc-amino ethyl bromide, n-boc-bromoethylamine, tert-butyl 2-bromoethyl carbamate, n-boc-2-bromoethylamine, tert-butyl 2-bromoethylcarbamate, 2-bromo-ethyl-carbamic acid tert-butyl ester, 2-tert-butoxycarbonylamino ethyl bromide, 2-boc-amino-ethyl bromide
PubChem CID 4103526
IUPAC Name tert-butyl N-(2-bromoethyl)carbamate
SMILES CC(C)(C)OC(=O)NCCBr

Specifications

Melting Point 33°C
Color White-Yellow
Quantity 1 g
Formula Weight 224.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(tert-Butoxycarbonylamino)ethyl Bromide
TSCA No
Recommended Storage Freezer