missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(tert-Butoxycarbonylamino)-1-ethanol 98.0+%, TCI America™
$100.62 - $288.59
Chemical Identifiers
| CAS | 26690-80-2 |
|---|---|
| Molecular Formula | C7H15NO3 |
| Molecular Weight (g/mol) | 161.201 |
| MDL Number | MFCD00056657 |
| InChI Key | GPTXCAZYUMDUMN-UHFFFAOYSA-N |
| Synonym | n-boc-ethanolamine, tert-butyl n-2-hydroxyethyl carbamate, boc-glycinol, tert-butyl 2-hydroxyethyl carbamate, n-tert-butoxycarbonyl ethanolamine, boc-ethanolamine, tert-butyl 2-hydroxyethylcarbamate, 2-tert-butoxycarbonylamino-1-ethanol, boc-aminoethanol, n-2-hydroxyethyl carbamic acid tert-butyl ester |
| PubChem CID | 2733206 |
| IUPAC Name | tert-butyl N-(2-hydroxyethyl)carbamate |
| SMILES | CC(C)(C)OC(=O)NCCO |
Chemical Identifiers
| 26690-80-2 | |
| 161.201 | |
| GPTXCAZYUMDUMN-UHFFFAOYSA-N | |
| 2733206 | |
| CC(C)(C)OC(=O)NCCO |
| C7H15NO3 | |
| MFCD00056657 | |
| n-boc-ethanolamine, tert-butyl n-2-hydroxyethyl carbamate, boc-glycinol, tert-butyl 2-hydroxyethyl carbamate, n-tert-butoxycarbonyl ethanolamine, boc-ethanolamine, tert-butyl 2-hydroxyethylcarbamate, 2-tert-butoxycarbonylamino-1-ethanol, boc-aminoethanol, n-2-hydroxyethyl carbamic acid tert-butyl ester | |
| tert-butyl N-(2-hydroxyethyl)carbamate |
Specifications
| 26690-80-2 | |
| Yellow | |
| MFCD00056657 | |
| GPTXCAZYUMDUMN-UHFFFAOYSA-N | |
| tert-butyl N-(2-hydroxyethyl)carbamate | |
| 161.201 | |
| 161.20 | |
| Liquid |
| 92°C | |
| C7H15NO3 | |
| n-boc-ethanolamine, tert-butyl n-2-hydroxyethyl carbamate, boc-glycinol, tert-butyl 2-hydroxyethyl carbamate, n-tert-butoxycarbonyl ethanolamine, boc-ethanolamine, tert-butyl 2-hydroxyethylcarbamate, 2-tert-butoxycarbonylamino-1-ethanol, boc-aminoethanol, n-2-hydroxyethyl carbamic acid tert-butyl ester | |
| CC(C)(C)OC(=O)NCCO | |
| 5 mL | |
| 2733206 | |
| ≥98.0% (GC) | |
| 2-(tert-Butoxycarbonylamino)-1-ethanol |
Safety and Handling
TSCA : Yes