missing translation for 'onlineSavingsMsg'
Learn More

2-(tert-Butoxycarbonyl)-2-azabicyclo[2.2.1]hept-5-ene 95.0+%, TCI America™

Catalog No. B48061G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B48061G Supplier TCI America Supplier No. B48061G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 188345-71-3
Molecular Formula C11H17NO2
Molecular Weight (g/mol) 195.262
MDL Number MFCD22416354
InChI Key ZATXHTCUZAWODK-UHFFFAOYSA-N
Synonym 2-boc-2-azabicyclo 2.2.1 hept-5-ene, tert-butyl 2-aza-bicyclo 2.2.1 hept-5-ene-2-carboxylate, tert-butyl 2-azabicyclo 2.2.1 hept-5-ene-2-carboxylate, 2-tert-butoxycarbonyl-2-azabicyclo 2.2.1 hept-5-ene, tert-butyl2-azabicyclo 2.2.1 hept-5-ene-2-carboxylate, ksc497c1n, tert-butyl 5-azabicyclo 2.2.1 hept-2-ene-5-carboxylate, 2-aza-bicyclo 2.2.1 hept-5-ene-2-carboxylic acid tert-butyl ester, 2-tert-butyloxycarbonyl-2-azabicyclo 2.2.1 hepta-5-ene, 2-azabicyclo 2.2.1 hept-5-ene-2-carboxylic acid tert-butyl ester
PubChem CID 15837921
IUPAC Name tert-butyl 5-azabicyclo[2.2.1]hept-2-ene-5-carboxylate
SMILES CC(C)(C)OC(=O)N1CC2CC1C=C2

Specifications

Color Yellow
Boiling Point 92°C
Quantity 1 g
Formula Weight 195.26
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(tert-Butoxycarbonyl)-2-azabicyclo[2.2.1]hept-5-ene