missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butoxy-6-chloropyridine, 97%

CAS: 547740-43-2 | C9H12ClNO | 185.651 g/mol

$43.60 - $561.37

Chemical Identifiers

CAS 547740-43-2
Molecular Formula C9H12ClNO
Molecular Weight (g/mol) 185.651
MDL Number MFCD08705263
InChI Key PLDRTFJRLUKTTA-UHFFFAOYSA-N
Synonym 2-tert-butoxy-6-chloropyridine, acmc-20anmt, 2-tert-butoxy-6-chloro-pyridine, pyridine, 2-chloro-6-1,1-dimethylethoxy
PubChem CID 11984350
IUPAC Name 2-chloro-6-[(2-methylpropan-2-yl)oxy]pyridine
SMILES CC(C)(C)OC1=NC(=CC=C1)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5508503
View Documents
Thermo Scientific Chemicals
H5508503
1 g
Each for $43.60
Only null left
 
AAH5508506
View Documents
Thermo Scientific Chemicals
H5508506
5 g
Each for $144.40
Only null left
 
AAH5508514
View Documents
Thermo Scientific Chemicals
H5508514
25 g
Each for $561.37
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

547740-43-2
185.651
PLDRTFJRLUKTTA-UHFFFAOYSA-N
11984350
CC(C)(C)OC1=NC(=CC=C1)Cl
C9H12ClNO
MFCD08705263
2-tert-butoxy-6-chloropyridine, acmc-20anmt, 2-tert-butoxy-6-chloro-pyridine, pyridine, 2-chloro-6-1,1-dimethylethoxy
2-chloro-6-[(2-methylpropan-2-yl)oxy]pyridine

Specifications

547740-43-2
1.076 g/mL
C9H12ClNO
1 g
PLDRTFJRLUKTTA-UHFFFAOYSA-N
2-chloro-6-[(2-methylpropan-2-yl)oxy]pyridine
11984350
97%
2-tert-Butoxy-6-chloropyridine
Colorless
78°C (172°F)
MFCD08705263
2-tert-butoxy-6-chloropyridine, acmc-20anmt, 2-tert-butoxy-6-chloro-pyridine, pyridine, 2-chloro-6-1,1-dimethylethoxy
CC(C)(C)OC1=NC(=CC=C1)Cl
185.651
185.65
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

P210-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c

H227-H302-H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only