missing translation for 'onlineSavingsMsg'
Learn More

2-Sulfobenzoic acid cyclic anhydride, 95%

CAS: 81-08-3 | C7H4O4S | 184.17 g/mol

$471.31 - $471.31

Chemical Identifiers

CAS 81-08-3
Molecular Formula C7H4O4S
Molecular Weight (g/mol) 184.17
MDL Number MFCD00005879
InChI Key NCYNKWQXFADUOZ-UHFFFAOYSA-N
Synonym 2-sulfobenzoic anhydride, 2-sulfobenzoic acid cyclic anhydride, sulfobenzoic anhydride, o-sulfobenzoic acid anhydride, 3h-2,1-benzoxathiol-3-one, 1,1-dioxide, 3h-benzo c 1,2 oxathiol-3-one 1,1-dioxide, unii-0swh68iq5w, o-sulfobenzoic acid, cyclic anhydride, 2,1-benzoxathiol-3-one-1,1-dioxide, 0swh68iq5w
PubChem CID 65729
IUPAC Name 1,1-dioxo-2,1$l^{6}-benzoxathiol-3-one
SMILES C1=CC=C2C(=C1)C(=O)OS2(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC184751000
SDS View Documents
Thermo Scientific Chemicals
184751000
100 g Glass bottle
Each for $471.31
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

81-08-3
184.17
NCYNKWQXFADUOZ-UHFFFAOYSA-N
65729
C1=CC=C2C(=C1)C(=O)OS2(=O)=O
C7H4O4S
MFCD00005879
2-sulfobenzoic anhydride, 2-sulfobenzoic acid cyclic anhydride, sulfobenzoic anhydride, o-sulfobenzoic acid anhydride, 3h-2,1-benzoxathiol-3-one, 1,1-dioxide, 3h-benzo c 1,2 oxathiol-3-one 1,1-dioxide, unii-0swh68iq5w, o-sulfobenzoic acid, cyclic anhydride, 2,1-benzoxathiol-3-one-1,1-dioxide, 0swh68iq5w
1,1-dioxo-2,1$l^{6}-benzoxathiol-3-one

Specifications

81-08-3
100.0
White to Beige
Authentic
Glass bottle
MFCD00005879
2-sulfobenzoic anhydride, 2-sulfobenzoic acid cyclic anhydride, sulfobenzoic anhydride, o-sulfobenzoic acid anhydride, 3h-2,1-benzoxathiol-3-one, 1,1-dioxide, 3h-benzo c 1,2 oxathiol-3-one 1,1-dioxide, unii-0swh68iq5w, o-sulfobenzoic acid, cyclic anhydride, 2,1-benzoxathiol-3-one-1,1-dioxide, 0swh68iq5w
C1=CC=C2C(=C1)C(=O)OS2(=O)=O
184.17
184.17
Technical
2-Sulfobenzoic acid cyclic anhydride, tech., 90%
94.0
116°C to 127°C
184°C to 186°C (18.0 mmHg)
95%
C7H4O4S
100 g
NCYNKWQXFADUOZ-UHFFFAOYSA-N
1,1-dioxo-2,1$l^{6}-benzoxathiol-3-one
65729
95%
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec

GHS Signal Word: Warning

EINECSNumber : 201-322-6

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only