missing translation for 'onlineSavingsMsg'
Learn More

2-Succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate, 98%

Catalog No. p-7046230
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1353806 5 g
AAL1353814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1353806 Supplier Thermo Scientific Chemicals Supplier No. L1353806
Only null left

CAS: 105832-38-0 | C9H16BF4N3O3 | 301.05 g/mol

2-Succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate (TSTU) acts as an efficient coupling agent through rapid activation of carboxylic acids to form N-succinimidyl active esters, which react with primary amines to form carboxamides. It is effective even in the presence of water. TSTU is selective towards amines in the presence of alcoholic group. It is employed as a coupling regent in the synthesis of peptides. With amine-protected alpha-amino acids, it brings about coupling without significant epimerization. It is useful for linking of sensor molecules with amino groups to amino-functionalized AFM (atomic force microscope) tips.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate (TSTU) acts as an efficient coupling agent through rapid activation of carboxylic acids to form N-succinimidyl active esters, which react with primary amines to form carboxamides. It is effective even in the presence of water. TSTU is selective towards amines in the presence of alcoholic group. It is employed as a coupling regent in the synthesis of peptides. With amine-protected alpha-amino acids, it brings about coupling without significant epimerization. It is useful for linking of sensor molecules with amino groups to amino-functionalized AFM (atomic force microscope) tips.

Solubility
Soluble in acetonitrile, DMF, chloroform, dichlorormethane and acetonitrile.

Notes
Moisture sensitive, Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 105832-38-0
Molecular Formula C9H16BF4N3O3
Molecular Weight (g/mol) 301.05
MDL Number MFCD00077875
InChI Key YEBLHMRPZHNTEK-UHFFFAOYSA-N
Synonym tstu, o-n-succinimidyl-1,1,3,3-tetramethyluronium tetrafluoroborate, 2-2,5-dioxopyrrolidin-1-yl-1,1,3,3-tetramethylisouronium tetrafluoroborate, 2-succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl-uronium tetrafluoroborate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl uronium bf4, pubchem12781
PubChem CID 9857522
IUPAC Name [dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C

Specifications

Melting Point 195°C to 200°C
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Soluble in acetonitrile,DMF,chloroform,dichlorormethane and acetonitrile.
Formula Weight 301.05
Percent Purity 98%
Chemical Name or Material 2-Succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only