missing translation for 'onlineSavingsMsg'
Learn More

2-sec-Butyl-3-methoxypyrazine 97.0+%, TCI America™

Catalog No. B29595G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29595G 5 g
B29591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29595G Supplier TCI America Supplier No. B29595G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24168-70-5
Molecular Formula C9H14N2O
Molecular Weight (g/mol) 166.224
MDL Number MFCD00006136
InChI Key QMQDJVIJVPEQHE-UHFFFAOYSA-N
Synonym 2-sec-butyl-3-methoxypyrazine, 2-methoxy-3-sec-butylpyrazine, 2-methoxy-3-1-methylpropyl pyrazine, pyrazine, 2-methoxy-3-1-methylpropyl, 2-methoxy-3-sec-butyl pyrazine, 3-sec-butyl-2-methoxypyrazine, pyrazine, 2-methoxy, 3-sec-butyl, 2-methoxy-3-1-methyl-propyl pyrazine
PubChem CID 520098
IUPAC Name 2-butan-2-yl-3-methoxypyrazine
SMILES CCC(C)C1=NC=CN=C1OC

Specifications

Color Yellow
Boiling Point 99°C
Quantity 5 g
Formula Weight 166.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-sec-Butyl-3-methoxypyrazine